CAS RN 10016-20-3
alpha-Cyclodextrin, 97+%
CAS: 10016-20-3 Molecular Formula: C36H60O30 Molecular Weight (g/mol): 972.846 MDL Number: MFCD00078207 InChI Key: HFHDHCJBZVLPGP-UHFFFAOYSA-N Synonym: cyclohexaamylose,cyclomaltohexose,.alpha.-cyclodextrin,schardinger alpha-dextrin,.alpha.-dextrin,.alpha.-cycloamylose,bdbm4,dextrin, .alpha.-cyclo,.alpha.-schardinger dextrin PubChem CID: 320760 SMILES: C(C1C2C(C(C(O1)OC3C(OC(C(C3O)O)OC4C(OC(C(C4O)O)OC5C(OC(C(C5O)O)OC6C(OC(C(C6O)O)OC7C(OC(O2)C(C7O)O)CO)CO)CO)CO)CO)O)O)O
α-Cyclodextrin, MP Biomedicals™
CAS: 10016-20-3 Molecular Formula: C36H60O30 Molecular Weight (g/mol): 972.846 InChI Key: HFHDHCJBZVLPGP-UHFFFAOYSA-N Synonym: cyclohexaamylose,cyclomaltohexose,.alpha.-cyclodextrin,schardinger alpha-dextrin,.alpha.-dextrin,.alpha.-cycloamylose,bdbm4,dextrin, .alpha.-cyclo,.alpha.-schardinger dextrin PubChem CID: 320760 SMILES: C(C1C2C(C(C(O1)OC3C(OC(C(C3O)O)OC4C(OC(C(C4O)O)OC5C(OC(C(C5O)O)OC6C(OC(C(C6O)O)OC7C(OC(O2)C(C7O)O)CO)CO)CO)CO)CO)O)O)O
R&D Systems™ a-Cyclodextrin
A Tocris product, part of R&D Systems. Cyclic complexing agent
alpha-Cyclodextrin, 98%, Spectrum™ Chemical
CAS: 10016-20-3 Molecular Formula: C36H60O30 Molecular Weight (g/mol): 972.85 InChI Key: HFHDHCJBZVLPGP-UHFFFAOYNA-N IUPAC Name: 5,10,15,20,25,30-hexakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29-dodecaoxaheptacyclo[26.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶]dotetracontane-31,32,33,34,35,36,37,38,39,40,41,42-dodecol SMILES: OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC1C(O)C2O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O
alpha-Cyclodextrin 98.0+%, TCI America™
CAS: 10016-20-3 Molecular Formula: C36H60O30 Molecular Weight (g/mol): 972.846 MDL Number: MFCD00078207 InChI Key: HFHDHCJBZVLPGP-UHFFFAOYSA-N Synonym: cyclohexaamylose,cyclomaltohexose,.alpha.-cyclodextrin,schardinger alpha-dextrin,.alpha.-dextrin,.alpha.-cycloamylose,bdbm4,dextrin, .alpha.-cyclo,.alpha.-schardinger dextrin PubChem CID: 320760 SMILES: C(C1C2C(C(C(O1)OC3C(OC(C(C3O)O)OC4C(OC(C(C4O)O)OC5C(OC(C(C5O)O)OC6C(OC(C(C6O)O)OC7C(OC(O2)C(C7O)O)CO)CO)CO)CO)CO)O)O)O
Sigma Aldrich L-Cysteine
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich (1R,3S)-(+)-Camphoric Acid
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Alpha-Cyclodextrin
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich (2-Hydroxypropyl)-Beta-Cyclodextrin
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Alpha-Cyclodextrin
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Alpha-Cyclodextrin
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Alpha-Cyclodextrin
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.