accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10020-43-6

IUPAC Name: 2-(octyloxy)ethan-1-ol
Synonyms: capryleth-4
Molecular Weight (g/mol): 174.28
Molecular Formula: C10H22O2
InChi Key: ZQCIMPBZCZUDJM-UHFFFAOYSA-N
SMILES: CCCCCCCCOCCO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results