Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Octyl-monooxyethylene, Thermo Scientific™

Catalog No. AC124710050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC124710050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC124710050 Supplier Thermo Scientific Chemicals Supplier No. 124710050

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10020-43-6
Molecular Formula C10H22O2
Molecular Weight (g/mol) 174.28
MDL Number MFCD00043070
InChI Key ZQCIMPBZCZUDJM-UHFFFAOYSA-N
Synonym 2-octyloxy ethanol, ethanol, 2-octyloxy, ethylene glycol monooctyl ether, ethyleneglycolmonooctylether, octyloxy ethanol, ethylene glycol mono-n-octyl ether, 2-octyloxyethan-1-ol, octyl alcohol, ethoxylated, n-octyl-polyoxyethylene, polyethylene glycol monooctyl ether
PubChem CID 82296
IUPAC Name 2-(octyloxy)ethan-1-ol
SMILES CCCCCCCCOCCO

Specifications

Density 0.88
Linear Formula CH3(CH2)7OCH2CH2OH
Quantity 5 g
Formula Weight 174.3
Chemical Name or Material n-Octyl-monooxyethylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.