accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2OH2OH2OH2OH2OH2OCoONOOONOO

CAS RN 10026-22-9

IUPAC Name: λ²-cobalt(2+) hexahydrate dinitrate
Synonyms: cobaltous nitrate hexahydrate
Molecular Weight (g/mol): 291.03
Molecular Formula: CoH12N2O12
InChi Key: QGUAJWGNOXCYJF-UHFFFAOYSA-N
SMILES: O.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Quantity 
  • (2)
  • (2)
  • (6)
  • (2)
  • (2)
  • (1)
  • (5)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (10)
Physical Form 
  • (3)
  • (4)
  • (8)
  • (3)
  • (11)
  • (1)
  • (3)
  • (2)
Grade 
  • (4)
  • (7)
  • (6)
  • (3)
  • (3)
  • (1)
  • (5)
  • (2)
  • (3)
Percent Purity 
  • (7)
  • (3)
  • (4)
  • (1)
  • (6)
  • (1)
  • (3)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (17)
  • (11)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (1)
  • (2)
  • (11)
  • (19)
  • (2)
  • (1)
  • (2)
  • (2)
  • (6)
  • (2)
  • (2)
  • (1)
  • (5)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (10)
  • (3)
  • (4)
  • (8)
  • (3)
  • (11)
  • (1)
  • (3)
  • (2)
  • (4)
  • (7)
  • (6)
  • (3)
  • (3)
  • (1)
  • (5)
  • (2)
  • (3)
  • (7)
  • (3)
  • (4)
  • (1)
  • (6)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)