Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobalt, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical™

Catalog No. SC193100 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SC193-100 100 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SC193-100

missing translation for 'validMainframeMsg'

For atomic absorption. (1mL = 1mg Co) Solute: Cobalt Nitrate Solvent: Dilute Nitric Acid

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10026-22-9 , 7732-18-5
Molecular Formula CoH12N2O12
Molecular Weight (g/mol) 291.03
MDL Number MFCD00149647
InChI Key QGUAJWGNOXCYJF-UHFFFAOYSA-N
PubChem CID 24821
ChEBI CHEBI:86214
IUPAC Name λ²-cobalt(2+) hexahydrate dinitrate
SMILES O.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Packaging Poly Bottle
Vapor Pressure 17mmHg at 20°C
Physical Form Liquid
Concentration 990 to 1010 ppm
Color Red
pH 2
Boiling Point 100°C
Quantity 100 mL
Grade Certified
Concentration or Composition (by Analyte or Components) 1000ppm ±1%
Chemical Name or Material Cobalt, Reference Standard Solution, 1000ppm ±1%
Show More Show Less
Safety handling Check Alert storage code Red
DOTInformation DOT Class 8, : Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.