accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1003-09-4

IUPAC Name: 2-bromothiophene
Synonyms: 2-bromothiophene
Molecular Weight (g/mol): 163.03
Molecular Formula: C4H3BrS
InChi Key: TUCRZHGAIRVWTI-UHFFFAOYSA-N
SMILES: BrC1=CC=CS1
Boiling Point 
  • (3)
  • (3)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (8)
  • (3)
Percent Purity 
  • (3)
  • (2)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (2)
  • (6)
  • (3)
  • (3)
  • (3)
  • (2)
  • (8)
  • (3)
  • (3)
  • (2)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (5)