CAS RN 10031-92-2
IUPAC Name:
ethyl non-2-ynoate
Synonyms:
ethyl non-2-ynoate
Molecular Weight (g/mol):
182.26
Molecular Formula:
C11H18O2
InChi Key:
BFZNMUGAZYAMTG-UHFFFAOYSA-N
SMILES:
CCCCCCC#CC(=O)OCC
1
–
1
of
1
results
1
Ethyl 2-Nonynoate, TCI America™
CAS: 10031-92-2 Molecular Formula: C11H18O2 Molecular Weight (g/mol): 182.263 MDL Number: MFCD00036553 InChI Key: BFZNMUGAZYAMTG-UHFFFAOYSA-N Synonym: ethyl 2-nonynoate,2-nonynoic acid, ethyl ester,ethyl-2-nonynoate,ethyl nonynoate,ethyl octynecarboxylate,2-nonynoic acid ethyl ester,ethyl octinecarbonate,unii-853ded009t,fema no. 2448,nonynoic acid, ethyl ester PubChem CID: 61451 IUPAC Name: ethyl non-2-ynoate SMILES: CCCCCCC#CC(=O)OCC