accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2H2NOHSOHOO

CAS RN 10034-93-2

IUPAC Name: hydrazine; sulfuric acid
Synonyms: hydrazine sulfate
Molecular Weight (g/mol): 130.12
Molecular Formula: H6N2O4S
InChi Key: ZGCHATBSUIJLRL-UHFFFAOYSA-N
SMILES: NN.OS(O)(=O)=O
Quantity 
  • (1)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (6)
  • (1)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (6)
  • (12)
Percent Purity 
  • (3)
  • (4)
  • (2)
  • (7)
  • (2)
Grade 
  • (7)
  • (4)
  • (1)
  • (4)
Physical Form 
  • (1)
  • (5)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (19)
  • (11)
  • (1)
  • (2)
  • (1)
  • (2)
  • (11)
  • (9)
  • (1)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (6)
  • (1)
  • (1)
  • (1)
  • (6)
  • (12)
  • (3)
  • (4)
  • (2)
  • (7)
  • (2)
  • (7)
  • (4)
  • (1)
  • (4)
  • (1)
  • (5)
  • (2)
  • (1)