Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrazine Sulfate (Powder/Certified ACS), Fisher Chemical

Catalog No. H320500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
H320-500 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. H320-500

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 10034-93-2
Molecular Formula H6N2O4S
Molecular Weight (g/mol) 130.118
MDL Number MFCD00044873
InChI Key ZGCHATBSUIJLRL-UHFFFAOYSA-N
Synonym hydrazine sulfate, hydrazine monosulfate, hydrazine, sulfate, hydrazine sulphate, hydrazinium sulfate, hydrazonium sulfate, hydrazine sulfate 1:1, segidrin, sehydrin, diamine sulfate
PubChem CID 24842
IUPAC Name hydrazine;sulfuric acid
SMILES NN.OS(=O)(=O)O

Specifications

Decomposition Information >254°C
Melting Point 254°C
Color White
Density 1.4g/mL
pH 1.5
Ignition Residue 0.05% max.
Assay Percent Range ≥99.0 %
Packaging Poly Bottle
Linear Formula NH2NH2·H2SO4
Quantity 500 g
UN Number 3260
Solubility Information 0.005% max. Insoluble Matter
Formula Weight 130.12
Percent Purity ≥99.0%
Grade Certified ACS
Physical Form Solid
Chemical Name or Material Hydrazine Sulfate
Show More Show Less
Safety handling Check Alert storage code Red
DOTInformation Class 8: Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.