accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1004-39-3

IUPAC Name: 4,6-diamino-1,2-dihydropyrimidine-2-thione
Synonyms: 4,6-diamino-1H-pyrimidine-2-thione
Molecular Weight (g/mol): 142.18
Molecular Formula: C4H6N4S
InChi Key: QCAWOHUJKPKOMD-UHFFFAOYSA-N
SMILES: NC1=CC(N)=NC(=S)N1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results