Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,6-Diamino-2-mercaptopyrimidine 95.0+%, TCI America™
SDP

Catalog No. D206325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D206325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D206325G Supplier TCI America Supplier No. D206325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1004-39-3
Molecular Formula C4H6N4S
Molecular Weight (g/mol) 142.18
MDL Number MFCD00006078
InChI Key QCAWOHUJKPKOMD-UHFFFAOYSA-N
Synonym 4,6-diamino-2-mercaptopyrimidine, 4,6-diaminopyrimidine-2-thiol, 2-mercapto-4,6-diaminopyrimidine, 2 1h-pyrimidinethione, 4,6-diamino, 4,6-diamino-2-thiopyrimidine, 4,6-diamino-2-pyrimidinethiol, 4,6-diaminopyrimidine-2 3h-thione, 4,6-diamino-2 1h-pyrimidinethione, qcawohujkpkomd-uhfffaoysa, hydrate
PubChem CID 1201482
IUPAC Name 4,6-diamino-1H-pyrimidine-2-thione
SMILES C1=C(NC(=S)N=C1N)N

Specifications

Color Yellow
Quantity 25 g
Formula Weight 142.18
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4,6-Diamino-2-mercaptopyrimidine
RTECSNumber MB7660000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.