accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1005201-24-0

IUPAC Name: 2-[3-(4-methanesulfinylphenyl)-1-benzofuran-5-yl]-5-methyl-1,3,4-oxadiazole
Synonyms: 2-[3-(4-methanesulfinylphenyl)-1-benzofuran-5-yl]-5-methyl-1,3,4-oxadiazole
Molecular Weight (g/mol): 338.38
Molecular Formula: C18H14N2O3S
InChi Key: RCKYSTKYIVULEK-UHFFFAOYNA-N
SMILES: CC1=NN=C(O1)C1=CC=C2OC=C(C2=C1)C1=CC=C(C=C1)S(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results