accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1005775-56-3

IUPAC Name: 4-tert-butyl-N-[6-(1H-imidazol-1-yl)imidazo[1,2-a]pyridin-2-yl]benzamide dihydrochloride
Synonyms: 4-tert-butyl-N-[6-(imidazol-1-yl)imidazo[1,2-a]pyridin-2-yl]benzamide dihydrochloride
Molecular Weight (g/mol): 432.35
Molecular Formula: C21H23Cl2N5O
InChi Key: IKKLFEDUYFZNBO-UHFFFAOYSA-N
SMILES: Cl.Cl.CC(C)(C)C1=CC=C(C=C1)C(=O)NC1=CN2C=C(C=CC2=N1)N1C=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results