accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFFCe

CAS RN 10060-10-3

IUPAC Name: λ⁴-cerium(4+) tetrafluoride
Synonyms: λ⁴-cerium(4+) tetrafluoride
Molecular Weight (g/mol): 216.11
Molecular Formula: CeF4
InChi Key: SWURHZJFFJEBEE-UHFFFAOYSA-J
SMILES: [F-].[F-].[F-].[F-].[Ce+4]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results