accessibility menu, dialog, popup

UserName

Viewing profile as user:
SOOOHONH(R)

CAS RN 1007840-62-1

IUPAC Name: (2R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-[(triphenylmethyl)sulfanyl]butanoic acid
Synonyms: (2R)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-4-[(triphenylmethyl)sulfanyl]butanoic acid
Molecular Weight (g/mol): 599.75
Molecular Formula: C38H33NO4S
InChi Key: FKBGJLDYRSFHBT-PGUFJCEWSA-N
SMILES: OC(=O)[C@@H](CCSC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)