accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1007847-70-2

IUPAC Name: 2-(oxan-4-yloxy)-6-(tributylstannyl)pyridine
Synonyms: 2-(oxan-4-yloxy)-6-(tributylstannyl)pyridine
Molecular Weight (g/mol): 468.27
Molecular Formula: C22H39NO2Sn
InChi Key: TYKSDTUCXMRYNR-UHFFFAOYSA-N
SMILES: CCCC[Sn](CCCC)(CCCC)C1=CC=CC(OC2CCOCC2)=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results