Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tributyl[6-(tetrahydropyran-4-yloxy)pyrid-2-yl]stannane, Thermo Scientific™

Catalog No. CC58241DE
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC58241DE 5 g Amber Glass Bottle
CC58241CB 250 mg Amber Glass Bottle
CC58241DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC58241DE Supplier Thermo Scientific Chemicals Supplier No. CC58241DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1007847-70-2
Molecular Formula C22H39NO2Sn
Molecular Weight (g/mol) 468.269
InChI Key TYKSDTUCXMRYNR-UHFFFAOYSA-N
Synonym 2-oxan-4-yloxy-6-tributylstannyl pyridine, tributyl 6-tetrahydropyran-4-yloxy pyrid-2-yl stannane, 2-oxan-4-yl oxy-6-tributylstannyl pyridine
PubChem CID 45594295
IUPAC Name tributyl-[6-(oxan-4-yloxy)pyridin-2-yl]stannane
SMILES CCCC[Sn](CCCC)(CCCC)C1=CC=CC(=N1)OC2CCOCC2

Specifications

Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 468.25
Percent Purity 90%
Chemical Name or Material tributyl[6-(tetrahydropyran-4-yloxy)pyrid-2-yl]stannane
Hazard Category GHS Signal Word: Danger
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.