accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 100828-16-8

IUPAC Name: 4-(3-phosphonopropyl)piperazine-2-carboxylic acid
Synonyms: 4-(3-phosphonopropyl)piperazine-2-carboxylic acid
Molecular Weight (g/mol): 252.21
Molecular Formula: C8H17N2O5P
InChi Key: CUVGUPIVTLGRGI-UHFFFAOYNA-N
SMILES: OC(=O)C1CN(CCCP(O)(O)=O)CCN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results