accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NOCH3

CAS RN 100848-70-2

IUPAC Name: 2-methoxy-4-methylpyridine
Synonyms: 2-methoxy-4-methylpyridine
Molecular Weight (g/mol): 123.16
Molecular Formula: C7H9NO
InChi Key: HGRXBKDKSYDWLD-UHFFFAOYSA-N
SMILES: COC1=NC=CC(C)=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results