CAS RN 1009373-58-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ 11beta-HSD1 Inhibitor, 10j
The 11beta-HSD1 Inhibitor, 10j, also referenced under CAS 1009373-58-3, controls the biological activity of 11beta-HSD1. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.