accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2OH2OH2OH2OH2OH2ONiOSOOO

CAS RN 10101-97-0

IUPAC Name: nickel(2+) hexahydrate sulfate
Synonyms: nickel sulfate hexahydrate nickel(II) sulfate hexahydrate nickelous sulfate hexahydrate
Molecular Weight (g/mol): 262.84
Molecular Formula: H12NiO10S
InChi Key: RRIWRJBSCGCBID-UHFFFAOYSA-L
SMILES: O.O.O.O.O.O.[Ni++].[O-]S([O-])(=O)=O
Molecular Weight (g/mol) 
  • (21)
  • (9)
  • (12)
Quantity 
  • (1)
  • (2)
  • (1)
  • (7)
  • (1)
  • (2)
  • (1)
  • (1)
  • (6)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (10)
  • (1)
Grade 
  • (4)
  • (8)
  • (7)
  • (4)
  • (2)
  • (1)
  • (2)
  • (4)
  • (2)
  • (1)
Physical Form 
  • (3)
  • (7)
  • (6)
  • (4)
  • (1)
  • (2)
  • (4)
  • (5)
Percent Purity 
  • (4)
  • (9)
  • (2)
  • (8)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (20)
  • (11)
  • (1)
  • (3)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (11)
  • (16)
  • (1)
  • (2)
  • (1)
  • (21)
  • (9)
  • (12)
  • (1)
  • (2)
  • (1)
  • (7)
  • (1)
  • (2)
  • (1)
  • (1)
  • (6)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (10)
  • (1)
  • (4)
  • (8)
  • (7)
  • (4)
  • (2)
  • (1)
  • (2)
  • (4)
  • (2)
  • (1)
  • (3)
  • (7)
  • (6)
  • (4)
  • (1)
  • (2)
  • (4)
  • (5)
  • (4)
  • (9)
  • (2)
  • (8)