Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel Standard, 1000 ppm Ni, Ricca Chemical
SDP

Catalog No. 530216 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
530216 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 530216 Supplier Ricca Chemical Company Supplier No. 530216
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 10101-97-0 , 7732-18-5
Molecular Formula H12NiO10S
Molecular Weight (g/mol) 262.839
InChI Key RRIWRJBSCGCBID-UHFFFAOYSA-L
PubChem CID 5284429
ChEBI CHEBI:53437
IUPAC Name nickel(2+);sulfate;hexahydrate
SMILES O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2]

Specifications

Packaging Natural Poly Bottle
Concentration or Composition Notes 1,000ppm Ni
CAS Min % 98.0, 0.44
CAS Max % 100.0, 0.46
Physical Form Liquid
Color Green
pH 4 to 5
Melting Point 0.0°C
Boiling Point 100°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type Analytical Standard
Quantity 500 mL
Grade Laboratory
Chemical Name or Material Nickel Standard Solution, Stock Solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.