Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel Sulfate Hexahydrate (Crystalline/Certified ACS), Fisher Chemical™

Catalog No. N73500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
N73-500 500 g Poly Bottle
N73-100 100 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N73-500

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 10101-97-0
Molecular Formula H12NiO10S
Molecular Weight (g/mol) 262.839
MDL Number MFCD00149813
InChI Key RRIWRJBSCGCBID-UHFFFAOYSA-L
Synonym nickel sulfate hexahydrate, nickel ii sulfate hexahydrate, nickel 2+ sulfate hexahydrate, nickel ii sulfate, nickelsulfatehexahydrate, dsstox_cid_928, acmc-1c7xm, dsstox_rid_75872, ni.so4.6h2o, dsstox_gsid_20928
PubChem CID 5284429
ChEBI CHEBI:53437
IUPAC Name nickel(2+);sulfate;hexahydrate
SMILES O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2]

Specifications

Boiling Point 103°C
Melting Point 53°C
Color Blue Green
pH 4 to 6
Physical Form Solid
Quantity 500 g
Formula Weight 262.85
Percent Purity 98 to 102%
Grade Certified ACS
Identification Pass Test
Packaging Poly Bottle
Decomposition Information >280°C
Density 2.07g/mL
Chemical Name or Material Nickel Sulfate Hexahydrate
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Cancer hazard. Harmful by inhalation and if swallowed. Irritating to eyes and skin. May cause allergic respiratory and skin reaction. May cause irritation of respiratory tract. May cause harm to the unborn child. Possible risks of irreversible effects. Danger of serious damage to health by prolonged exposure. Very toxic to aquatic organisms
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.