accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaOMoOOOHOHHOH

CAS RN 10102-40-6

IUPAC Name: disodium dioxomolybdenumbis(olate) dihydrate
Synonyms: sodium molybdate dihydrate
Molecular Weight (g/mol): 241.96
Molecular Formula: H4MoNa2O6
InChi Key: FDEIWTXVNPKYDL-UHFFFAOYSA-N
SMILES: O.O.[Na+].[Na+].[O-][Mo]([O-])(=O)=O
Grade 
  • (3)
  • (3)
  • (9)
  • (1)
  • (2)
  • (4)
  • (7)
  • (7)
  • (2)
Molecular Weight (g/mol) 
  • (12)
  • (26)
Quantity 
  • (1)
  • (2)
  • (1)
  • (5)
  • (2)
  • (2)
  • (5)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (9)
Percent Purity 
  • (7)
  • (2)
  • (4)
  • (9)
  • (1)
  • (2)
  • (8)
Physical Form 
  • (2)
  • (4)
  • (3)
  • (12)
  • (1)
  • (3)
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (14)
  • (12)
  • (2)
  • (1)
  • (2)
  • (5)
  • (1)
  • (1)
  • (4)
  • (12)
  • (12)
  • (2)
  • (3)
  • (3)
  • (9)
  • (1)
  • (2)
  • (4)
  • (7)
  • (7)
  • (2)
  • (12)
  • (26)
  • (1)
  • (2)
  • (1)
  • (5)
  • (2)
  • (2)
  • (5)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (9)
  • (7)
  • (2)
  • (4)
  • (9)
  • (1)
  • (2)
  • (8)
  • (2)
  • (4)
  • (3)
  • (12)
  • (1)
  • (3)
  • (6)
  • (4)