Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Molybdate Dihydrate, EMPLURA™, MilliporeSigma™

Catalog No. M1065241000 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Plastic Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1065241000 1 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1065241000 Supplier MilliporeSigma Supplier No. 1065241000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity sodium molybdate dihydrate for use in technical applications.

Recommended Storage

+5°C to +30°C.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10102-40-6
Molecular Formula H4MoNa2O6
Molecular Weight (g/mol) 241.96
MDL Number MFCD00149170
InChI Key FDEIWTXVNPKYDL-UHFFFAOYSA-N
Synonym sodium molybdate dihydrate, sodium molybdate vi dihydrate, disodium molybdate dihydrate, unii-8f2sxi1704, na2moo4.2h2o, disodium dihydrate molybdate, molybdic acid sodium salt dihydrate, molybdic acid, disodium salt, dihydrate, sodium molybdate 2 hydrate, dipotassium dihydrate molybdate
PubChem CID 16211258
ChEBI CHEBI:75213
IUPAC Name disodium dioxomolybdenumbis(olate) dihydrate
SMILES O.O.[Na+].[Na+].[O-][Mo]([O-])(=O)=O

Specifications

Melting Point 687°C (Elimination of water of crystallization)
Color White
pH 9 to 10 (840g/L, H2O, 20°C)
Physical Form Crystals
Quantity 1 kg
Solubility Information 840g/L
Formula Weight 241.95
Percent Purity 98.0 to 103.0%
Grade Not Available
Packaging Plastic Bottle
Density 2.71 g/3 (22 °C) (for the dihydrate)
Chemical Name or Material Sodium Molybdate Dihydrate
Show More Show Less
EINECSNumber 231-551-7
RTECSNumber QA5085000
Recommended Storage +5°C to +30°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.