accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOHNHOCl

CAS RN 101072-54-2

IUPAC Name: 2-(2-chloroacetamido)butanoic acid
Synonyms: 2-(2-chloroacetamido)butanoic acid
Molecular Weight (g/mol): 179.6
Molecular Formula: C6H10ClNO3
InChi Key: CLYGVTIFEIIGIX-UHFFFAOYNA-N
SMILES: CCC(NC(=O)CCl)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results