accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOOOONHN(S)

CAS RN 101214-43-1

IUPAC Name: 2,5-dioxopyrrolidin-1-yl (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-phenylpropanoate
Synonyms: 2,5-dioxopyrrolidin-1-yl (2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-phenylpropanoate
Molecular Weight (g/mol): 484.51
Molecular Formula: C28H24N2O6
InChi Key: VLXHZQQUTCVLGU-DEOSSOPVSA-N
SMILES: O=C(N[C@@H](CC1=CC=CC=C1)C(=O)ON1C(=O)CCC1=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)