accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH3CH3COHPOHOHO

CAS RN 10138-93-9

IUPAC Name: phosphoric acid; triethylamine
Synonyms: phosphoric acid; triethylamine
Molecular Weight (g/mol): 199.19
Molecular Formula: C6H18NO4P
InChi Key: UNXNGGMLCSMSLH-UHFFFAOYSA-N
SMILES: OP(O)(O)=O.CCN(CC)CC
Quantity 
  • (1)
  • (1)
pH 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results