accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOH

CAS RN 101385-90-4

IUPAC Name: 1-benzylpyrrolidin-3-ol
Synonyms: 1-benzylpyrrolidin-3-ol
Molecular Weight (g/mol): 177.25
Molecular Formula: C11H15NO
InChi Key: YQMXOIAIYXXXEE-UHFFFAOYNA-N
SMILES: OC1CCN(CC2=CC=CC=C2)C1
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)