accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OCH3CH3

CAS RN 10143-60-9

IUPAC Name: 2-ethyl-1-[(2-ethylhexyl)oxy]hexane
Synonyms: hexane, 1,1'-oxybis 2-ethyl-
Molecular Weight (g/mol): 242.45
Molecular Formula: C16H34O
InChi Key: YHCCCMIWRBJYHG-UHFFFAOYNA-N
SMILES: CCCCC(CC)COCC(CC)CCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results