Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(2-ethylhexyl) Ether 85.0+%, TCI America™
SDP

Catalog No. D102125ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D102125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D102125ML Supplier TCI America Supplier No. D102125ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 10143-60-9
Molecular Formula C16H34O
Molecular Weight (g/mol) 242.447
MDL Number MFCD00059427
InChI Key YHCCCMIWRBJYHG-UHFFFAOYSA-N
Synonym Di(2-ethylhexyl) Ether, Dioctyl Ether, 2-Ethylhexyl Ether, Octyl Ether
PubChem CID 25010
IUPAC Name 3-(2-ethylhexoxymethyl)heptane
SMILES CCCCC(CC)COCC(CC)CCCC

Specifications

Boiling Point 126°C
Quantity 25 mL
Formula Weight 242.45
Percent Purity ≥85.0% (GC)
Physical Form Liquid
Chemical Name or Material Bis(2-ethylhexyl) Ether
EINECSNumber (2)-0366
RTECSNumber KN3175000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.