accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3OOHOCH3Br

CAS RN 101460-22-4

IUPAC Name: 3-bromo-2,5,6-trimethoxybenzoic acid
Synonyms: 3-bromo-2,5,6-trimethoxybenzoic acid
Molecular Weight (g/mol): 291.1
Molecular Formula: C10H11BrO5
InChi Key: YHDRAWVCUZZYCU-UHFFFAOYSA-N
SMILES: COC1=CC(Br)=C(OC)C(C(O)=O)=C1OC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results