Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-2,5,6-trimethoxybenzoic Acid 97.0+%, TCI America™
SDP

Catalog No. B17401G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B17401G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B17401G Supplier TCI America Supplier No. B17401G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 101460-22-4
Molecular Formula C10H11BrO5
Molecular Weight (g/mol) 291.097
MDL Number MFCD00191620
InChI Key YHDRAWVCUZZYCU-UHFFFAOYSA-N
PubChem CID 625925
IUPAC Name 3-bromo-2,5,6-trimethoxybenzoic acid
SMILES COC1=CC(=C(C(=C1OC)C(=O)O)OC)Br

Specifications

Melting Point 96°C
Quantity 1 g
Formula Weight 291.10
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-2,5,6-trimethoxybenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.