accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1014691-61-2

IUPAC Name: methyl 3-{[2-methoxy-4-(phenylamino)phenyl]sulfamoyl}thiophene-2-carboxylate
Synonyms: methyl 3-{[2-methoxy-4-(phenylamino)phenyl]sulfamoyl}thiophene-2-carboxylate
Molecular Weight (g/mol): 418.48
Molecular Formula: C19H18N2O5S2
InChi Key: NDFKBGWLUHKMFY-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(C=CS1)S(=O)(=O)NC1=C(OC)C=C(NC2=CC=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results