accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1014979-56-6

IUPAC Name: (5-ethyl-2-methylpyridin-1-ium-1-yl)-λ¹-boranuidyl
Synonyms: (5-ethyl-2-methylpyridin-1-ium-1-yl)-λ¹-boranuidyl
Molecular Weight (g/mol): 131.99
Molecular Formula: C8H11BN
InChi Key: NHLPUHQNHDWXBY-UHFFFAOYSA-N
SMILES: [B-][N+]1=C(C)C=CC(CC)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results