accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNNHNNHOHN

CAS RN 101622-51-9

IUPAC Name: 2-{[6-(benzylamino)-9-methyl-9H-purin-2-yl]amino}ethan-1-ol
Synonyms: olomoucine
Molecular Weight (g/mol): 298.35
Molecular Formula: C15H18N6O
InChi Key: GTVPOLSIJWJJNY-UHFFFAOYSA-N
SMILES: CN1C=NC2=C(NCC3=CC=CC=C3)N=C(NCCO)N=C12
Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results