accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CO(S)(S)OH(S)OHOHN(S)CH3O(R)O(S)(S)HO(R)H3CO(R)(S)(S)OH(R)CH3(R)OH(R)CH3CH3H3CH3CNOCH3(R)OPHOOHOCH3H3CNNCH3H3C(E)(E)(E)(E)(Z)

CAS RN 101932-71-2

IUPAC Name: {[(2R,3R,5S,7R,8R,9S)-2-[(1S,3S,4R,5R,6R,7E,9E,11E,13Z)-14-cyano-3,5-dihydroxy-1-methoxy-4,6,8,9,13-pentamethyltetradeca-7,9,11,13-tetraen-1-yl]-7-[(2E)-3-{2-[(2S)-4-[(2S,3S,4S)-4-(dimethylamino)-2,3-dihydroxy-5-methoxypentanamido]butan-2-yl]-1,3-oxazol-4-yl}prop-2-en-1-yl]-9-hydroxy-4,4,8-trimethyl-1,6-dioxaspiro[4.5]decan-3-yl]oxy}phosphonic acid
Synonyms: [(2R,3R,5S,7R,8R,9S)-2-[(1S,3S,4R,5R,6R,7E,9E,11E,13Z)-14-cyano-3,5-dihydroxy-1-methoxy-4,6,8,9,13-pentamethyltetradeca-7,9,11,13-tetraen-1-yl]-7-[(2E)-3-{2-[(2S)-4-[(2S,3S,4S)-4-(dimethylamino)-2,3-dihydroxy-5-methoxypentanamido]butan-2-yl]-1,3-oxazol-4-yl}prop-2-en-1-yl]-9-hydroxy-4,4,8-trimethyl-1,6-dioxaspiro[4.5]decan-3-yl]oxyphosphonic acid
Molecular Weight (g/mol): 1009.19
Molecular Formula: C50H81N4O15P
InChi Key: FKAWLXNLHHIHLA-QJLNTFFJSA-N
SMILES: COC[C@@H]([C@H](O)[C@H](O)C(=O)NCC[C@H](C)C1=NC(\C=C\C[C@H]2O[C@@]3(C[C@H](O)[C@H]2C)O[C@H]([C@H](C[C@H](O)[C@@H](C)[C@H](O)[C@H](C)\C=C(/C)\C(\C)=C\C=C\C(\C)=C/C#N)OC)[C@H](OP(O)(O)=O)C3(C)C)=CO1)N(C)C
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)