Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Calyculin A, 98+%

Catalog No. AAJ61952EXG
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61952EXG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ61952EXG Supplier Thermo Scientific™ Supplier No. J61952EXG

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 μg
Description White
Content And Storage -30°C to -10°C
For Use With (Application) Very potent inhibitor (Ki = ca. 0.1 nM) with high specificity for the PP-1 and PP-2 classes of protein serine/threonine phosphatase
Assay 98+%
CAS 101932-71-2
Molecular Formula C50H81N4O15P
Formula Weight 1009.17
Form Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.