CAS RN 1019582-23-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-(4-Fluoro-2-nitrophenyl)-4-piperidone, 97%, Thermo Scientific™
CAS: 1019582-23-0