accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 102-11-4

IUPAC Name: benzyl(methyl)[2-(methylamino)ethyl]amine
Synonyms: benzyl(methyl)[2-(methylamino)ethyl]amine
Molecular Weight (g/mol): 178.28
Molecular Formula: C11H18N2
InChi Key: PVFIKWLTVKSXED-UHFFFAOYSA-N
SMILES: CNCCN(C)CC1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results