Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzyl-N,N'-dimethylethylenediamine 98.0+%, TCI America™
SDP

Catalog No. B32465G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32465G 5 g
B324625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B32465G Supplier TCI America Supplier No. B32465G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 102-11-4
Molecular Formula C11H18N2
Molecular Weight (g/mol) 178.28
MDL Number MFCD00014856
InChI Key PVFIKWLTVKSXED-UHFFFAOYSA-N
PubChem CID 66016
IUPAC Name benzyl(methyl)[2-(methylamino)ethyl]amine
SMILES CNCCN(C)CC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 74°C
Quantity 5 g
Formula Weight 178.28
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N-Benzyl-N,N′-dimethylethylenediamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.