accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONHH2N

CAS RN 102-28-3

IUPAC Name: N-(3-aminophenyl)acetamide
Synonyms: acetamide, N-(3-aminophenyl)-
Molecular Weight (g/mol): 150.18
Molecular Formula: C8H10N2O
InChi Key: PEMGGJDINLGTON-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=CC(N)=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)