Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3'-Aminoacetanilide, 97%, Thermo Scientific™

Catalog No. AAH6062930
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6062930 250 g
AAH6062918 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6062930 Supplier Thermo Scientific Chemicals Supplier No. H6062930

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3′-Aminoacetanilide is mainly used as reaction intermediates dyes and disperse dyes.

Solubility
Soluble in water 1-5 g/100 mL at 24°C.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-28-3
Molecular Formula C8H10N2O
Molecular Weight (g/mol) 150.18
MDL Number MFCD00025229
InChI Key PEMGGJDINLGTON-UHFFFAOYSA-N
Synonym n-3-aminophenyl acetamide, 3'-aminoacetanilide, 3-aminoacetanilide, m-aminoacetanilide, acetamide, n-3-aminophenyl, 3-acetamidoaniline, m-acetaminoaniline, m-acetylamino aniline, 3-aminoacetanilid, 3-amino-n-acetylaniline
PubChem CID 7604
IUPAC Name N-(3-aminophenyl)acetamide
SMILES CC(=O)NC1=CC=CC(N)=C1

Specifications

Melting Point 86°C to 88°C
Quantity 250 g
Solubility Information Soluble in water 1-5g/100 mL at 24°C.
Formula Weight 150.18
Percent Purity 97%
Chemical Name or Material 3'-Aminoacetanilide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 203-021-5
RTECSNumber AD8050000
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.