Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3'-Aminoacetanilide 98.0+%, TCI America™
SDP

Catalog No. A153325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A153325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A153325G Supplier TCI America Supplier No. A153325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 102-28-3
Molecular Formula C8H10N2O
Molecular Weight (g/mol) 150.18
MDL Number MFCD00025229
InChI Key PEMGGJDINLGTON-UHFFFAOYSA-N
Synonym n-3-aminophenyl acetamide, 3'-aminoacetanilide, 3-aminoacetanilide, m-aminoacetanilide, acetamide, n-3-aminophenyl, 3-acetamidoaniline, m-acetaminoaniline, m-acetylamino aniline, 3-aminoacetanilid, 3-amino-n-acetylaniline
PubChem CID 7604
IUPAC Name N-(3-aminophenyl)acetamide
SMILES CC(=O)NC1=CC=CC(N)=C1

Specifications

Melting Point 88°C
Quantity 25 g
Formula Weight 150.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3′-Aminoacetanilide
EINECSNumber (3)-0244
RTECSNumber AD8050000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.