accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH2H3C

CAS RN 102-48-7

IUPAC Name: 1-(3,4-dimethylphenyl)methanamine
Synonyms: 1-(3,4-dimethylphenyl)methanamine
Molecular Weight (g/mol): 135.21
Molecular Formula: C9H13N
InChi Key: PXNRCZQMDSDSHJ-UHFFFAOYSA-N
SMILES: CC1=CC=C(CN)C=C1C
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results