accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NONCH3NHNO

CAS RN 102146-07-6

IUPAC Name: 8-cyclopentyl-1,3-dipropyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
Synonyms: dipropylcyclopentylxanthine
Molecular Weight (g/mol): 304.39
Molecular Formula: C16H24N4O2
InChi Key: FFBDFADSZUINTG-UHFFFAOYSA-N
SMILES: CCCN1C2=C(NC(=N2)C2CCCC2)C(=O)N(CCC)C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)