CAS RN 1021868-90-5
IUPAC Name:
3-(propan-2-yl)bicyclo[2.2.1]heptan-2-amine hydrochloride
Synonyms:
3-isopropylbicyclo[2.2.1]heptan-2-amine hydrochloride
Molecular Weight (g/mol):
189.73
Molecular Formula:
C10H20ClN
InChi Key:
KSTGYMXUFHCTSM-UHFFFAOYNA-N
SMILES:
Cl.CC(C)C1C2CCC(C2)C1N
1
–
1
of
1
results
1
Tocris Bioscience™ AGN 192403 hydrochloride
CAS: 1021868-90-5 Molecular Formula: C10H20ClN