accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 102601-60-5

H3CNNNNH2CH3CH2CCH3CH3OOHOHOClCo

CAS RN 102601-60-5

IUPAC Name: 15,19-bis(2-carboxyethyl)-4,9-diethenyl-5,10,14,20-tetramethyl-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1(20),2,4,6(24),7,9,11(23),12,14,16,18-undecaene-21,22-diide; chlorocobaltbis(ylium)
Synonyms: 15,19-bis(2-carboxyethyl)-4,9-diethenyl-5,10,14,20-tetramethyl-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1(20),2,4,6(24),7,9,11(23),12,14,16,18-undecaene-21,22-diide; chlorocobaltbis(ylium)
Molecular Weight (g/mol): 655.04
Molecular Formula: C34H32ClCoN4O4
InChi Key: WWNXKQFLDNSNDN-UHFFFAOYSA-K
SMILES: Cl[Co++].CC1=C(CCC(O)=O)C2=CC3=C(CCC(O)=O)C(C)=C([N-]3)C=C3N=C(C=C4N=C(C=C1[N-]2)C(C)=C4C=C)C(C)=C3C=C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)