accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OPNH2OSH3C

CAS RN 10265-92-6

IUPAC Name: [methoxy(methylsulfanyl)phosphoryl]amine
Synonyms: tamaron sniper methamidophos chevron 9006 chevron ortho 9006 patrole ortho monitor MTD methyl phosphoramidothioate epa pesticide chemical code 101201
Molecular Weight (g/mol): 141.12
Molecular Formula: C2H8NO2PS
InChi Key: NNKVPIKMPCQWCG-UHFFFAOYNA-N
SMILES: COP(N)(=O)SC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results