CAS RN 102683-14-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Crypthophane E 95%, Thermo Scientific™
CAS: 102683-14-7 Molecular Formula: C57H60O12 Synonym: Crypthophane collet